Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Ethyl acetate ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
    
 
undefined
 
 
Ethyl acetate ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.5 %
  • Density (D 20/20):
    0.898 - 0.902
  • Density (D 25/25):
    0.894 - 0.898
  • Refractive index (n 20/D):
    1.371 - 1.373
  • Non-volatile matter:
    Max. 0.003 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.005 %
  • Ethanol (GC):
    Max. 0.1 %
  • Methyl compounds (as Acetic acid methyl ester):
    Max. 0.01 %
  • Reaction against H₂SO₄:
    complying
  • Related substances (GC):
    Max. 0.2 %
  • Chromatographic purity:
    complying
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying
  • Refractive Index:
    n20/D 1.371-1.373
  • Vapor Pressure:
    73 mmHg ( 20 °C)
  • Explosion Limit:
    2.2-11.5 %, 38 °F

 

 

Frequently Bought Together