Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Acetone ≥99.5% ACS, Riedel-de Haën™
    
 
undefined
 
 
Acetone ≥99.5% ACS, Riedel-de Haën™
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Identity (IR):
    complying
  • Assay according to ACS (GC):
    Min. 99.5 %
  • Solubility in water:
    complying
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.5 %
  • Titrable Acid:
    Max. 0.0003 meq/g
  • Titrable Base:
    Max. 0.0006 meq/g
  • KMnO₄ reducing substances:
    complying
  • Aldehydes (as HCHO):
    Max. 0.002 %
  • Methanol (GC):
    Max. 0.05 %
  • 2-propanol (GC):
    Max. 0.05 %
  • APHA:
    Max. 10
  • Vapor Pressure:
    184 mmHg ( 20 °C)
  • Vapor Density:
    2 (vs air)

 

 

Frequently Bought Together