Order Entry
Belgium
ContactUsLinkComponent
Acetone ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Acetone ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
   Sustainable Options Available
Certificates
undefined
Acetone ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Vapor Density:
    2 (vs air)
  • Refractive Index:
    n20/D 1.359 (lit.)

Specification Test Results

Related Information

Frequently Bought Together