Order Entry
Belgium
ContactUsLinkComponent
Calcium chloride hexahydrate 97-103%, meets analytical specification of Ph. Eur., E509 (max. 0.0001% Al), Fluka™
Irritant
Calcium chloride hexahydrate 97-103%, meets analytical specification of Ph. Eur., E509 (max. 0.0001% Al), Fluka™
Certificates
Irritant
Calcium chloride hexahydrate 97-103%, meets analytical specification of Ph. Eur., E509 (max. 0.0001% Al), Fluka™
Certificates

Specification Test Results

  • Transition Temp:
    solidification point 28-30 °C
  • Identity:
    complying
  • Assay:
    97.0 - 103.0 %
  • Solidification range:
    28 - 30 °C
  • Acid. or alk. react. impurities:
    complying
  • Aluminium (Al):
    Max. 1 ppm
  • Arsenic (As):
    Max. 1 ppm
  • Barium (Ba):
    complying
  • Iron (Fe):
    Max. 5 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Magnesium and alkali metals:
    Max. 0.3 %
  • Fluoride (F):
    Max. 40 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying

Specification Test Results

Related Information

Frequently Bought Together