Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™
Certificates
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (5 %, 25 °C):
    8.7 - 9.3
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

Related Information

Frequently Bought Together