Biodiesel calibration standards
: Restek
The biodiesel component is methyl soyate.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
- CAS No.:621-71-6
- Formula:C₃₃H₆₂O₆
- Description:Tricaprin (1,2,3-tricaprinoylglycerol)
Frequently Bought Together