ASTM precision thermometers, stem type
Supplier: Ludwig Schneider
Jump to:
About this item
These precision thermometers are available with DAkks calibration certificate.
- Round, white back capillary form
Blue filling, stem type
: Packed individualy in a square plastic sleeve.
8 Options Available Below
Product Details & Documents
These precision thermometers are available with DAkks calibration certificate.
- Round, white back capillary form
Blue filling, stem type
: Packed individualy in a square plastic sleeve.
Product Family Options
Product Information
- RangeDivisionLengthFillingImmersion depthØAvailabilityPrice
- SCNL1202005S/04/3PASTM S5C, with 3 calibration pointsASTM S5C, with 3 calibration points−38...+50 °C1 °C260 mmBlue108 mm6 - 8 mmLog in for availability1 * 1 ST388,00 €
- SCNL1202062S/04/3PASTM S62C, with 3 calibration pointsASTM S62C, with 3 calibration points−38...+2 °C0,1 °C401 mmBlueTotal6 - 8 mmLog in for availability1 * 1 ST485,00 €
- SCNL1202012S/04/3PASTM S12C, with 3 calibration pointsASTM S12C, with 3 calibration points−20...+102 °C0,2 °C435 mmBlueTotal6 - 8 mmLog in for availability1 * 1 ST484,00 €
- SCNL1202015S/04/3PASTM S15C, with 3 calibration pointsASTM S15C, with 3 calibration points−2...+80 °C0,2 °C415 mmBlueTotal6 - 8 mmLog in for availability1 * 1 ST422,00 €
- SCNL1202091S/04/3PASTM S91C, with 3 calibration pointsASTM S91C, with 3 calibration points+20...+50 °C0,1 °C390 mmBlue76 mm6 - 8 mmLog in for availability1 * 1 ST433,00 €
- SCNL1202064S/04/5PASTM S64C, with 5 calibration pointsASTM S64C, with 5 calibration points+25...+55 °C0,1 °C401 mmBlueTotal6 - 8 mmLog in for availability1 * 1 ST665,00 €
- SCNL1202018S/04/3PASTM S18C, with 3 calibration pointsASTM S18C, with 3 calibration points+34...+42 °C0,1 °C300 mmBlueTotal6 - 8 mmLog in for availability1 * 1 ST411,00 €
- SCNL1202120S/04/3PASTM S120C, With 3 calibration pointsASTM S120C, With 3 calibration points+38,6...+41,4 °C0,05 °C300 mmBlueTotal6 - 8 mmLog in for availability1 * 1 ST492,00 €