Order Entry
Belgium
ContactUsLinkComponent
Barium chloride 0.1 mol/l (0.2N) in aqueous solution, AVS TITRINORM volumetric solution
Barium chloride 0.1 mol/l (0.2N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 85496.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Barium chloride 0.1 mol/l (0.2N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 85496.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0,0998 -> 0,1002 mol/l

Specification Test Results

About this item

 :
 :208.24 g/mol
Storage Temperature:Ambient
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445