Order Entry
Belgium
ContactUsLinkComponent
Cerium(IV) sulphate 0.25 mol/l (0.25 N) in 0.25 N sulphuric acid, AVS TITRINORM volumetric solution
Cerium(IV) sulphate 0.25 mol/l (0.25 N) in 0.25 N sulphuric acid, AVS TITRINORM volumetric solution
  85804.290
 :  
undefined
Cerium(IV) sulphate 0.25 mol/l (0.25 N) in 0.25 N sulphuric acid, AVS TITRINORM volumetric solution
  85804.290
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Titer (20°C; real value 0.2 % accuracy):
    0.2495 -> 0.2505 mol/l

 

 

 :
 :332.24 g/mol
 :Ambient
 :13590-82-4
 :Ce(SO₄)₂
 :MFCD00148852