Order Entry
Belgium
ContactUsLinkComponent
Potassium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Potassium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 85809.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 85809.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :74.55 g/mol
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360