Order Entry
Belgium
ContactUsLinkComponent
 
Mouse Recombinant IL-7 (Animal free) (from E. coli)
 
undefined
Mouse Recombinant IL-7 (Animal free) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: 2E8 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Gene ID:
    P10168
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    IL-7|Lymphopoietin 1 (LP-1)|pre-B cell Factor|Interleukin 7
  • Protein/peptide name:
    IL7
  • Purity:
    ≥95%
  • Molecular weight:
    15 kDa
  • Sequence:
    MECHIKDKEG KAYESVLMIS IDELDKMTGT DSNCPNNEPN FFRKHVCDDT KEAAFLNRAA RKLKQFLKMN ISEEFNVHLL TVSQGTQTLV NCTSKEEKNV KEQKKNDACF LKRLLREIKT CWNKILKGSI
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together