Chlordane, toxaphene solutions
: Restek
Chlordane and toxaphene standards may exhibit lot-to-lot variation.
- Certified reference materials
- Manufactured and QC-tested in ISO-accredited labs
- CAS No.:57-74-9
- Formula:C₁₀H₆Cl₈
- Description:Chlordane
Frequently Bought Together