Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 8 mol/l (8 N) in aqueous solution, AVS TITRINORM volumetric solution
Corrosive
Sodium hydroxide 8 mol/l (8 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 89745.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 8 mol/l (8 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 89745.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :40 g/mol
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548