Diesel fuel #2 (ASTM Class 5 Accelerant)
Supplier: Restek
Certificates
About this item
These solutions are prepared from a single-source (one-refinery) product.
- Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs
The weathered materials indicate the percent weight loss from the original material.
Frequently Bought Together