Order Entry
Belgium
ContactUsLinkComponent
Acetic acid 2%, VOLUSOL®
Acetic acid 2%, VOLUSOL®
Catalog # 86139.310
Supplier:  VWR Chemicals
CAS Number:  
undefined
Acetic acid 2%, VOLUSOL®
Catalog # 86139.310
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    1.9 --> 2.1 %

Specification Test Results

About this item

 :
 :60.05 g/mol
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152