Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 10 mol/l 10 N, AVS TITRINORM
Corrosive
Sodium hydroxide 10 mol/l 10 N, AVS TITRINORM
Catalog # 5041.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 10 mol/l 10 N, AVS TITRINORM
Catalog # 5041.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer:
    9.5 - 10.5 mol/l

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548