Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium bromide
Potassium bromide 0.1 M in aqueous solution
Potassium bromide 0.1 M in aqueous solution
85808.290
:
VWR Chemicals
:
Potassium bromide 0.1 M in aqueous solution
85808.290
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
119 g/mol
:
7758-02-3
:
KBr
:
MFCD00011358