Order Entry
Belgium
ContactUsLinkComponent
Multi-element ICP-MS calibrating solution - 4 components Ca ; Mg ; K ; Na in 2% HNO₃
Multi-element ICP-MS calibrating solution - 4 components Ca ; Mg ; K ; Na in 2% HNO₃
Catalog # ACSDMS91C81K2NL1
Supplier:  CPACHEM
CAS Number:  
Multi-element ICP-MS calibrating solution - 4 components Ca ; Mg ; K ; Na in 2% HNO₃
Catalog # ACSDMS91C81K2NL1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L each
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element calibrating solutions Ca ; Mg ; K ; Na

Specifications

About this item

ICP-MS calibration standard - 4 components