Order Entry
Belgium
ContactUsLinkComponent
Magnesium chloride hexahydrate 98.0-101.0% BP, Ph. Eur., pharma grade
Magnesium chloride hexahydrate 98.0-101.0% BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates
Magnesium chloride hexahydrate 98.0-101.0% BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (Compl.):
    98.0 - 101.0%
  • Identity according to Pharmacopoeias:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Acidity or Alkalinity:
    Passes Test
  • Ammonium (NH₄):
    0.005%
  • Bromide (Br):
    0.05%
  • Phosphate (PO₄):
    0.003%
  • Sulfate (SO₄):
    0.01%
  • Residual solvents (Ph.Eur.):
    Passes Test
  • Water (H₂O):
    51.0 - 55.0%
  • Heavy metals (as Pb):
    0.001%
  • Al:
    0.0001%
  • As:
    0.00015%
  • Ca:
    0.1%
  • Fe:
    0.001%
  • Cd, (Class 1 - ICH Q3D):
    0,5 ppm
  • Pb, (Class 1 - ICH Q3D):
    0,5 ppm
  • As, (Class 1 - ICH Q3D):
    1,5 ppm
  • Hg, (Class 1 - ICH Q3D):
    1,5 ppm
  • Co, (Class 2A - ICH Q3D):
    5 ppm
  • V, (Class 2A - ICH Q3D):
    10 ppm
  • Ni, (Class 2A - ICH Q3D):
    20 ppm
  • Tl, (Class 2B - ICH Q3D):
    0,8 ppm
  • Au, (Class 2B - ICH Q3D):
    10 ppm
  • Pd, (Class 2B - ICH Q3D):
    10 ppm
  • Ir, (Class 2B - ICH Q3D):
    10 ppm
  • Os, (Class 2B - ICH Q3D):
    10 ppm
  • Rh, (Class 2B - ICH Q3D):
    10 ppm
  • Ru, (Class 2B - ICH Q3D):
    10 ppm
  • Se, (Class 2B - ICH Q3D):
    15 ppm
  • Ag, (Class 2B - ICH Q3D):
    15 ppm
  • Pt, (Class 2B - ICH Q3D):
    10 ppm
  • Li, (Class 3 - ICH Q3D):
    55 ppm
  • Sb, (Class 3 - ICH Q3D):
    120 ppm
  • Ba, (Class 3 - ICH Q3D):
    140 ppm
  • Mo, (Class 3 - ICH Q3D):
    25 ppm
  • Cu, (Class 3 - ICH Q3D):
    250 ppm
  • Sn, (Class 3 - ICH Q3D):
    600 ppm
  • Cr, (Class 3 - ICH Q3D):
    25 ppm

Specification Test Results

Frequently Bought Together