Order Entry
Belgium
ContactUsLinkComponent
Analytical standard, Dinotefuran, reference material, Honeywell Fluka™
Analytical standard, Dinotefuran, reference material, Honeywell Fluka™
  HONCR1344-50MG
 :  
Analytical standard, Dinotefuran, reference material, Honeywell Fluka™
  HONCR1344-50MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    165252-70-0
  • Description:
    Dinotefuran
  • Melting point:
    101…105 °C
  • Formula:
    C₇H₁₄N₄O₃
  • Storage temperature:
    Ambient
  • Description:
    Dinotefuran

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results