Analytical standard, Flumetsulam, reference material, Honeywell Fluka™
HONCR1315-50MG
:
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:98967-40-9
- Description:Flumetsulam
- Melting point:251…253 °C
- Formula:C₁₂H₉F₂N₅O₂S
- Storage temperature:Ambient
- Description:Flumetsulam
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results