Analytical standard, Cyprodinil-(phenyl-¹³C₆), reference material, Honeywell Fluka™
HONCR1303-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1773496-63-1
- Description:Cyprodinil-(phenyl-¹³C₆)
- Melting point:63…67 °C
- Formula:¹³C₆C₈H₁₅N₃
- Storage temperature:Ambient
- Description:Cyprodinil-(phenyl-¹³C₆)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results