Analytical standard, Paraquat diiodide-(dimethyl-d₆), reference material, Honeywell Fluka™
HONCR1281-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- Description:Paraquat diiodide-(dimethyl-d₆)
- Melting point:343 °C
- Formula:C₁₂D₆H₈I₂N₂
- Storage temperature:2…8 °C
- Description:Paraquat diiodide-(dimethyl-d₆)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results