Order Entry
Belgium
ContactUsLinkComponent
 
Analytical standard, R1266 Ternidazole HCL, reference material, Honeywell Fluka™
  HONCR1266-10MG
 :  
Analytical standard, R1266 Ternidazole HCL, reference material, Honeywell Fluka™
  HONCR1266-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    70028-95-4
  • Description:
    R1266 Ternidazole HCL
  • Melting point:
    121…126 °C
  • Formula:
    C₇H₁₁N₃O₃ · HCl
  • Storage temperature:
    Ambient
  • Description:
    R1266 Ternidazole HCL

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results