Order Entry
Belgium
ContactUsLinkComponent
 
Analytical standard, Buparvaquone, reference material, Honeywell Fluka™
Catalog # HONCR1255-25MG
CAS Number:  
Analytical standard, Buparvaquone, reference material, Honeywell Fluka™
Catalog # HONCR1255-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    88426-33-9
  • Description:
    Buparvaquone
  • Melting point:
    177…185 °C
  • Formula:
    C₂₁H₂₆O₃
  • Storage temperature:
    2…8 °C
  • Description:
    Buparvaquone

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results