Order Entry
Belgium
ContactUsLinkComponent
Analytical standard, erythro-Isoxsuprine hydrochloride, reference material, Honeywell Fluka™
Analytical standard, erythro-Isoxsuprine hydrochloride, reference material, Honeywell Fluka™
  HONCR1229-10MG
 :  
Analytical standard, erythro-Isoxsuprine hydrochloride, reference material, Honeywell Fluka™
  HONCR1229-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    579-56-6
  • Description:
    erythro-Isoxsuprine hydrochloride
  • Melting point:
    219…223 °C
  • Formula:
    C₁₈H₂₃NO₃ · HCl
  • Storage temperature:
    Ambient
  • Description:
    erythro-Isoxsuprine hydrochloride

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results