Order Entry
Belgium
ContactUsLinkComponent
 
Analytical standard, Butyl 4-hydroxybenzoate-(ring-¹³C₆), reference material, Honeywell Fluka™
  HONCR1179-10MG
 :  
Analytical standard, Butyl 4-hydroxybenzoate-(ring-¹³C₆), reference material, Honeywell Fluka™
  HONCR1179-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1416711-53-9
  • Description:
    Butyl 4-hydroxybenzoate-(ring-¹³C₆)
  • Melting point:
    67…72 °C
  • Formula:
    ¹³C₆C₅H₁₄O₃
  • Storage temperature:
    2…8 °C
  • Description:
    Butyl 4-hydroxybenzoate-(ring-¹³C₆)
  • Boiling point:
    156…157 °C

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results