Analytical standard, Voriconazole, reference material, Honeywell Fluka™
Catalog # HONCR1174-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:137234-62-9
- Description:Voriconazole
- Environmentally Preferable:
- Melting point:128…134 °C
- Formula:C₁₆H₁₄F₃N₅O
- Storage temperature:Ambient
- Description:Voriconazole
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results