Analytical standard, Cylindrospermopsin solution, reference material, Honeywell Fluka™
HONCR1087-1ML
:
- Pack type:Glass Bottle
- Pk:1 mL
- Standards:Analytical standard, single component solution
- CAS No.:143545-90-8
- Description:Cylindrospermopsin solution, 10 μg/ml in water
- Formula:C₁₅H₂₁N₅O₇S
- Storage temperature:–20°C
- Description:Cylindrospermopsin solution
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results