Analytical standard, Azimsulfuron, reference material, Honeywell Fluka™
HONCR1074-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:120162-55-2
- Description:Azimsulfuron
- Formula:C₁₃H₁₆N₁₀O₅S
- Storage temperature:Ambient
- Description:Azimsulfuron
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results