Order Entry
Belgium
ContactUsLinkComponent
Analytical standard, Etoricoxib, reference material, Honeywell Fluka™
Analytical standard, Etoricoxib, reference material, Honeywell Fluka™
Catalog # HONCR1063-10MG
CAS Number:  
Analytical standard, Etoricoxib, reference material, Honeywell Fluka™
Catalog # HONCR1063-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    202409-33-4
  • Description:
    Etoricoxib
  • Melting point:
    134…135 °C
  • Formula:
    C₁₈H₁₅ClN₂O₂S
  • Storage temperature:
    2…8 °C
  • Description:
    Etoricoxib

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results