Order Entry
Belgium
ContactUsLinkComponent
Analytical standard, Ofloxacin-d₃, reference material, Honeywell Fluka™
Analytical standard, Ofloxacin-d₃, reference material, Honeywell Fluka™
Catalog # HONCR1008-10MG
CAS Number:  
Analytical standard, Ofloxacin-d₃, reference material, Honeywell Fluka™
Catalog # HONCR1008-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1173147-91-5
  • Description:
    Ofloxacin-d₃
  • Melting point:
    269…270 °C
  • Formula:
    C₁₈D₃H₁₇FN₃O₄
  • Storage temperature:
    Ambient
  • Description:
    Ofloxacin-d₃

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results