Order Entry
Belgium
ContactUsLinkComponent
 
Analytical standard, Tolfenamic acid-(benzoic ring-¹³C₆), reference material, Honeywell Fluka™
  HONCR1001-10MG
 :  
Analytical standard, Tolfenamic acid-(benzoic ring-¹³C₆), reference material, Honeywell Fluka™
  HONCR1001-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1420043-61-3
  • Description:
    Tolfenamic acid-(benzoic ring-¹³C₆)
  • Environmentally Preferable:
  • Melting point:
    208…213 °C
  • Formula:
    ¹³C₆C₈H₁₂ClNO₂
  • Storage temperature:
    Ambient
  • Description:
    Tolfenamic acid-(benzoic ring-¹³C₆)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results