Potassium Standard for IC, TraceCERT®, Supelco®
Catalog # 53337-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O/tr HNO₃
- Pk:100 mL
- Environmentally Preferable:
- M-Clarity quality level:MQ300
- Element:Potassium
- Symbol:K
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034