About this item
The Defender 5000 series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, stainless steel frame and a stainless steel (single range models) or aluminium load cell (dual range models). The front mounted D52XW indicator is 304 stainless steel with sand blasted surface treatment, which offers protection to IP 68.
- Large backlit display and alpha-numeric keypad with function keys
- Indicator guides user with on-screen prompts
- Multiple application modes and high storage capacity for greater flexibility
- Multiple connectivity options for convenient data transfer and storage
Equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management, and multiple connectivity options - the Defender 5000 is designed to simplify demanding industrial and commercial applications.
The series features common industrial modes and supports a 50000 item library and 1000 user profiles which can be downloaded/uploaded for easy set-up (with SD card.) Also equipped with GLP/GMP output capability with time/date for result traceability.
Weighing results can also be stored in Alibi or regular format, addressing all your data storage needs. Data output via built-in RS232 or optional additional interfaces including second RS232/RS485/USB device, Ethernet, Bluetooth/WiFi, analogue output or discrete I/O with relays, the Defender 5000 indicator can also be used for systems including PLCs.
Results can be read easily on the Defender 5000's large transflective dot matrix display, even in bright sunlight. The SmarText 3.0 interface will guide you through all of the Defender 5000's advanced features. With five soft keys, SmartText 3.0 makes an advanced scale very simple to use, and reduces training time for operators.
Featuring most common applications including percent weighing, parts counting with APW optimisation, checkweighing or checkcounting and dynamic weighing or display hold, the Defender 5000 meets the weighing and measurement needs of practically every industrial application or manufacturing process. Many application modes eliminate the need for long and complicated manual calculations. GMP/GLP with time/date allows for traceability of results.