Order Entry
Belgium
ContactUsLinkComponent
D(-)-Luciferin potassium salt ≥99.5% endotoxin-free
D(-)-Luciferin potassium salt ≥99.5% endotoxin-free
  OZBILK10000
 :  
undefined
D(-)-Luciferin potassium salt ≥99.5% endotoxin-free
  OZBILK10000
 :  

 

  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

 

 

 : (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate
 :
 :318.41 g/mol
 :115144-35-9
 :C₁₁H₇KN₂O₃S₂
 :MFCD00044928