ContactUsLinkComponent
Digital hygrometer for Secador desiccators, SP Bel-Art
Digital hygrometer for Secador desiccators, SP Bel-Art
Catalog # BELAF420701400
undefined
Digital hygrometer for Secador desiccators, SP Bel-Art
Catalog # BELAF420701400
Specifications
  • Description:
    Digital hygrometer
  • Environmentally Preferable:
  • Accuracy:
    ±3%rH (%rH values from 20 to 80%)
    ±7%rH (%rH values from 10 to 20%)
  • Pk:
    1
Specifications
About this item
These replacement digital hygrometer fits any Secador® desiccator equipped with a door mounted hygrometer.
  • Display is refreshed every ten seconds
  • The long term drift in the accuracy of the hygrometer is 0,5% rH per year
This hygrometer displays a two-digit number followed by % rH for relative humidity (rH) values between 10 and 80% rH (% rH values below 10% will show '<10% rH' and % rH values above 80% will show '>80% rH'). It is powered by a CR-2032 battery which provides approximately 1 year battery life.