HPLC columns, sugar analysis, NUCLEOSIL® Carbohydrate
Supplier: MACHEREY-NAGEL
Certificates
About this item
Silica-based phase for HPLC separation of mono- and disaccharides.
- Robust spherical silica gel
- Amino modification
- RP separation of mono- and disaccharides
NUCLEOSIL® Carbohydrate is recommended for easy separations of mono- and dissacharides under reversed phase conditions.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications
- Catalog No:
- MANA720905.40
- MANA721170.30
- USP code:
- L8
- L8