ContactUsLinkComponent
Jar, PP, with screw cap, SP Bel-Art
Jar, PP, with screw cap, SP Bel-Art
Catalog # BELAH179100000
undefined
Jar, PP, with screw cap, SP Bel-Art
Catalog # BELAH179100000
Specifications
  • Ext.Ø:
    58 mm
  • Capacity:
    118 ml
  • Height:
    66,5 mm
  • Pk:
    12
Specifications
About this item
White PP jar with screw cap in an assortment of sizes and is used for storage of liquid or dry stock, or for use with samples.
Screw cap with PE foam lining and are not autoclavable.