Orders LinkContactUsLinkComponent
Storage drums
Supplier:   Sampling Systems
Storage drums
Supplier:  Sampling Systems
Certificates
Storage drums
7 Options Available Below
Product Details & Documents
These drums are used for the storage and processing of powders and liquids.
  • Open top pharmaceutical grade drums with lids
  • Made of 316L stainless steel body and lid
  • Supplied with side handles
The robust, hygienic design means they are suitable for the most demanding applications. These drums are finished to a bright polish finish with all the welds ground and polished.
 : Supplied with lid, clamp and O-ring.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 1 L
    155 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 519,00 €
    519,00 €
    1 * 1 ST
       519,00 €
  • 2 L
    155 mm
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 537,00 €
    537,00 €
    1 * 1 ST
       537,00 €
  • 3 L
    192 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 549,00 €
    549,00 €
    1 * 1 ST
       549,00 €
  • 5 L
    196 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 627,00 €
    627,00 €
    1 * 1 ST
       627,00 €
  • 10 L
    223 mm
    270 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 788,00 €
    788,00 €
    1 * 1 ST
       788,00 €
  • 15 L
    298 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 849,00 €
    849,00 €
    1 * 1 ST
       849,00 €
  • 50 L
    393 mm
    435 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 190,00 €
    1 190,00 €
    1 * 1 ST
       1 190,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Silicone gasket (FDA acceptable), 196 mm int.Ø of drum
    Silicone gasket (FDA acceptable), 196 mm int.Ø of drum
    SAMPA430-196
    Log in for availability
    1 * 1 ST - 40,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Drum lid, 316L SS, 395 mm int.Ø of drum
    Drum lid, 316L SS, 395 mm int.Ø of drum
    SAMPA429-395
    Log in for availability
    1 * 1 ST - 353,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone gasket (FDA acceptable), 300 mm int.Ø of drum
    Silicone gasket (FDA acceptable), 300 mm int.Ø of drum
    SAMPA430-300
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Drum lid, 316L SS, 470 mm int.Ø of drum
    Drum lid, 316L SS, 470 mm int.Ø of drum
    SAMPA429-470
    Log in for availability
    1 * 1 ST - 362,00 €