Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Amarell
Precision thermometers
Supplier:  Amarell
Certificates
Precision thermometers
18 Options Available Below
Product Details & Documents
These thermometers are suitable for government verification or calibration.
  • With ring at top
  • Durable pigment
  • Solid stem
Packaging: Packed in a transparent square plastic case.
Product Family Options
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • −10...+100 °C
    ±0,5 %
    300 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,30 €
    49,30 €
    1 * 1 ST
       49,30 €
  • −10...+100 °C
    ±1,0 %
    305 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,90 €
    38,90 €
    1 * 1 ST
       38,90 €
  • −10...+150 °C
    ±1,0 %
    305 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,20 €
    43,20 €
    1 * 1 ST
       43,20 €
  • −10...+250 °C
    ±1,0 %
    350 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,40 €
    72,40 €
    1 * 1 ST
       72,40 €
  • −10...+150 °C
    ±0,5 %
    350 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • −10...+360 °C
    ±1,0 %
    380 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €
  • −5...+100 °C
    ±0,2 %
    420 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,40 €
    65,40 €
    1 * 1 ST
       65,40 €
  • −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 78,80 €
    78,80 €
    1 * 1 ST
       78,80 €
With calibration certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • −5...+50 °C
    ±0,2 %
    350 mm
    Blue special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,60 €
    95,60 €
    1 * 1 ST
       95,60 €
  • 0...+50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 130,00 €
    130,00 €
    1 * 1 ST
       130,00 €
With DKD certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • −10...+100 °C
    ±0,5 %
    300 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 241,00 €
    241,00 €
    1 * 1 ST
       241,00 €
  • −5...+100 °C
    ±0,2 %
    420 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
With verification certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • −10...+100 °C
    ±0,5 %
    300 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 328,00 €
    328,00 €
    1 * 1 ST
       328,00 €
  • −100...+100 °C
    ±1,0 %
    300 mm
    Toluene
    8 - 9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 426,00 €
    426,00 €
    1 * 1 ST
       426,00 €
  • 0...+100 °C
    ±1,0 %
    305 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 304,00 €
    304,00 €
    1 * 1 ST
       304,00 €
  • 0...+360 °C
    ±1,0 %
    380 mm
    Gallium
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 564,00 €
    564,00 €
    1 * 1 ST
       564,00 €
With work certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • −5...+100 °C
    ±0,2 %
    420 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST