Order Entry
Belgium
ContactUsLinkComponent
Trifluoroacetic acid ≥99.0%, LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Trifluoroacetic acid ≥99.0%, LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Certificates
undefined
Trifluoroacetic acid ≥99.0%, LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99 %
  • Suitability for the LC-MS:
    complying
  • Water (Karl Fischer):
    Max. 0.05 %
  • Absorbance at 320 nm:
    Max. 0.025
  • Absorbance at 300 nm:
    Max. 0.03
  • Absorbance at 290 nm:
    Max. 0.04
  • Absorbance 280 nm:
    Max. 0.05
  • Absorbance at 270 nm:
    Max. 0.1
  • Absorbance at 260 nm:
    Max. 0.9
  • Silver (Ag):
    Max. 0.02 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Barium (Ba):
    Max. 0.05 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.2 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.05 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.3 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.02 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Manganese (Mn):
    Max. 0.05 ppm
  • Molybdenum (Mo):
    Max. 0.02 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.05 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.02 ppm
  • Thallium (Tl):
    Max. 0.05 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • Sulfite (SO₃):
    Max. 10 ppm
  • Ammonium (NH₄):
    Max. 10 ppm

Specification Test Results

Related Information

Frequently Bought Together