Orders LinkContactUsLinkComponent
DIN adapters for electrodes
DIN adapters for electrodes
Supplier:   Mettler - Toledo
DIN adapters for electrodes
Supplier:  Mettler - Toledo
Certificates
DIN adapters for electrodes
2 Options Available Below
Product Details & Documents
These adapters will allow the LE electrodes with BNC to connect to meters with DIN.
Product Family Options
Product Information
  • Availability
    Price
  • METT30403890
    DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427
    DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,60 €
    31,60 €
    1 * 1 ST
       31,60 €
  • METT30499900
    DIN 19262 adapter T kit including temperature for LE438 / LE410
    DIN 19262 adapter T kit including temperature for LE438 / LE410
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,20 €
    46,20 €
    1 * 1 ST
       46,20 €