Orders LinkContactUsLinkComponent
Partial immerstion thermometers
Partial immerstion thermometers
Supplier:   Amarell
Partial immerstion thermometers
Supplier:  Amarell
Certificates
Partial immerstion thermometers
4 Options Available Below
Product Details & Documents
Thermometers are permanently marked.
  • Celsius scale
  • Partial immersion
  • Yellow back
Immersion depth: 76 mm.
Product Family Options
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Availability
    Price
  • −10...+50 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,00 €
    8,00 €
    1 * 1 ST
       8,00 €
  • −10...+110 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,90 €
    7,90 €
    1 * 1 ST
       7,90 €
  • −10...+150 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,30 €
    8,30 €
    1 * 1 ST
       8,30 €
  • −10...+50 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,80 €
    8,80 €
    1 * 1 ST
       8,80 €