Orders LinkContactUsLinkComponent
LED illuminator
LED illuminator
LED illuminator
Catalog # ZEIS4239049902000
Supplier:   ZEISS
LED illuminator
Catalog # ZEIS4239049902000
Supplier:  ZEISS
LED illuminator
LED illuminator
Price
677,71 €
Log in for availability
1 * 1 ST
Product Details & Documents
LED illuminator for transmitted light for direct attachment to the condenser carrier.
Contains sliders with diffusion disk with 10° divergence angle for visual observation and with 80° divergence angle for documentation. Slider can also be used as shutter against interfering autofluorescence of the LEDs in epi-fluorescence applications.
Specifications
  • Description:
    LED illuminator