Order Entry
Belgium
ContactUsLinkComponent
Rappaport Vassiliadis broth, modified, suitable for microbiology, NutriSelect® Plus
Rappaport Vassiliadis broth, modified, suitable for microbiology, NutriSelect® Plus
Catalog # SIAL17173-500G
Supplier:  Merck
CAS Number:  
Rappaport Vassiliadis broth, modified, suitable for microbiology, NutriSelect® Plus
Catalog # SIAL17173-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 g
  • Description:
    Rappaport vassiliadis broth, modified

Specifications

About this item

Used as a selective enrichment medium for the isolation of Salmonella species from food and environmental specimens.

  • pH: 5,2 ±0,2 (25 °C)
  • Limited shelf life

Contains papaic digest of soyabean meal 5 g/1110 ml; Sodium chloride 8 g/1110 ml; Monopotassium phosphate 1,6 g/1110 ml; Magnesium chloride anhydrous 18,7 g/L (is equivalent to 40 g/1110 ml Magnesium chloride hexahydrate) and malachite green 0,04 g/1110 ml.