Orders LinkContactUsLinkComponent
Pilot Remote software
Pilot Remote software
Pilot Remote software, Huber temperature control units with Pilot ONE
Catalog # HUBE10645
Supplier:   HUBER
Pilot Remote software
Catalog # HUBE10645
Supplier:  HUBER
Pilot Remote software
Pilot Remote software
Pilot Remote software, Huber temperature control units with Pilot ONE
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The Pilot Remote software enables the complete remote control of temperature control units with Pilot ONE through a Windows PC.
  • Remote control of any number of temperature control units with Pilot ONE
  • Secure Ethernet connection with authentication
  • Compatible with Microsoft Windows 7 / 8 / 10 (32 / 64 bit)
The user display of the Pilot ONE is equally displayed on the PC, this means identical operation on PC and temperature control unit. The communication takes place via Ethernet network connectivity with a secure authentication and encoding.
Delivery information: Supplied with hardware dongle (for any Windows PC).
Specifications
  • Description:
    Pilot Remote software
  • For:
    Huber temperature control units with Pilot ONE