Orders LinkContactUsLinkComponent
Cleanroom shelf
Cleanroom shelf
Cleanroom mini shelf, solid, Stainless steel 304, 450×600×260 mm
Catalog # PALBMB06045-S
Supplier:   Palbam
Cleanroom shelf
Catalog # PALBMB06045-S
Supplier:  Palbam
Cleanroom shelf
Cleanroom shelf
Cleanroom mini shelf, solid, Stainless steel 304, 450×600×260 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Wall mounted mini shelf can be fixed anywhere. Fabrication material is stainless steel 304 with electropolished finish.
Specifications
  • Description:
    Cleanroom mini shelf, solid
  • Material:
    Stainless steel 304
  • W×D×H:
    450×600×260 mm