Order Entry
Belgium
ContactUsLinkComponent
Activity test strip, ExitusPlus™
Activity test strip, ExitusPlus™
Catalog # A9411.0025
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Activity test strip, ExitusPlus™
Catalog # A9411.0025
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    25
  • Description:
    Activity tests, ExitusPlus™, 25 strips/pack
  • Storage temperature:
    Ambient

Specifications

About this item

DNA/RNA-ExitusPlus™ activity test is a quick and convenient tool to test the activity of PanReac AppliChem solutions for nucleic acid decontamination DNA/RNA-ExitusPlus™(A7089), DNA/RNA-ExitusPlus™ IF (A7409) and RNase-ExitusPlus™ (A7153). The test strips are very sensitive and designed to detect the redox reaction of DNA/RNA-ExitusPlus™ solutions.

  • Very sensitive
  • Calibrated to nucleic acid degradation
  • Can detect ageing of solutions

Use the strips to test DNA/RNA-ExitusPlus™ if there are any concerns about the activity of the solution, e.g. after the recommended re-test date or after repeated use in decontamination protocols. Follow the recommendations given in the colour scale. DNA/RNA-ExitusPlus™ solutions that give results within the sufficient efficacy range may be further used for nucleic acid decontamination.

Instructions for Use:
1. Use one strip for one sample. Soak the strip into decontamination solution for 10 seconds.
2. Remove the test strip, shake well to remove excess solution.
3. Wait for 30 sec. to develop the full color.
4. Match the color developed on the strip to the color scale.

Caution: Store strips at room temperature.