Ring lights, EasyLED
The EasyLED product line integrates advanced LED illumination with control functions in a single compact device. It is the ideal standard illumination system for routine inspections and education.
- Powerful illuminance
- Compact and ergonomic
- Integrated controller
- Mountable direct to the microscope objective
- International clip plug system
The EasyLED ring light offers homogeneous and shadow-free illumination in a robust metal housing and well designed heat sink that enables maximum brightness and long lifetime of 50000 hours. The multi LED light head offers an attractive alternative to conventional cold light sources with fiber optics.
The EasyLED ring light plus has the additional advantage of controllable segments, which enables new contrasting methods. The integrated 'jog dial' element allows an easy and intuitive switching between the different illumination modes as well as rotation of the segments in both directions.
The EasyLED product line integrates advanced LED illumination with control functions in a single compact device. It is the ideal standard illumination system for routine inspections and education.
- Powerful illuminance
- Compact and ergonomic
- Integrated controller
- Mountable direct to the microscope objective
- International clip plug system
The EasyLED ring light offers homogeneous and shadow-free illumination in a robust metal housing and well designed heat sink that enables maximum brightness and long lifetime of 50000 hours. The multi LED light head offers an attractive alternative to conventional cold light sources with fiber optics.
The EasyLED ring light plus has the additional advantage of controllable segments, which enables new contrasting methods. The integrated 'jog dial' element allows an easy and intuitive switching between the different illumination modes as well as rotation of the segments in both directions.
Documents
AT-ce_easyled_rl AT-ce_easyled_rlplus TDS EasyLED Ringlights TDS Ringlights TDS System Diagram for EasyLED Series
Datasheet
AT-Video_EasyLED_Ringlight_PLUS_INT EasyLED Ringlight PLUS LED Cold Light Sources Ring Lights, EasyLED
Videos
EasyLED Series Standard Illumination for Stereo Microscopy
Product Literature
SCOC600300
Images 360
Product Family Options
- Description
- EasyLED ring light system, FWD 55 - 135 mm, max. illuminance 90 klx (at 75 mm FWD), Øi 66 mm Øe 114 mm1 * 1 ST550,00 €
- EasyLED ring light plus system, FWD 50 - 130 mm, max. illuminance 140 klx (at 75 mm FWD), Øi 66 mm Øe 114 mm1 * 1 ST600,00 €